Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Halogenated Pesticides and Herbicides Blend, SPEX CertiPrep™
SDP

Catalog No. 5511PH Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5511-PH 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5511-PH Supplier SPEX CertiPrep Supplier No. 5511PH

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Alachlor15972-60-82000μg/mL
Atrazine1912-24-92000μg/mL
Bladex21725-46-22000μg/mL
Bromacil314-40-92000μg/mL
Endrin72-20-82000μg/mL
Endrin aldehyde7421-93-42000μg/mL
Endrin ketone53494-70-52000μg/mL
gamma-BHC58-89-92000μg/mL
Heptachlor76-44-82000μg/mL
Heptachlor epoxide (Isomer B)1024-57-32000μg/mL
Hexachlorobenzene118-74-12000μg/mL
Hexachlorocyclopentadiene77-47-42000μg/mL
Methoxychlor72-43-52000μg/mL
Metolachlor51218-45-22000μg/mL
Metribuzin21087-64-92000μg/mL
Trifluralin1582-09-82000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Halogenated Pesticides and Herbicides Mix
Packaging 2mL amber prescored glass ampule
Name Note USEPA Method 551.1
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Methods For US EPA Method 551, 551.1
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solvent or Matrix Acetone
Quantity 1 mL

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.