Learn More
TARGETMOL CHEMICALS INC HALOPEMIDE 5MG
SDP
Catalog No. 502577353
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Halopemide is a potent inhibitor of PLD (IC50 = 220 and 310 nM for human PLD1 and PLD2). Halopemid is an antagonist of dopamine receptors. Halopemid can be used in psychotropic research. purity: 96%
Catalog No. 50-257-7353 Supplier TARGETMOL CHEMICALS INC Supplier No. T154615MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Halopemide is a potent inhibitor of PLD (IC50 = 220 and 310 nM for human PLD1 and PLD2). Halopemid is an antagonist of dopamine receptors. Halopemid can be used in psychotropic research. purity: 96%