Learn More
Medchemexpress LLC HY-119093 5mg Medchemexpress, Halopemide CAS:59831-65-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1190935MG
Encompass_Preferred
Medchemexpress, HY-119093 5mg Halopemide CAS:59831-65-1 Halopemide is a potent phospholipase D (PLD) inhibitor, with IC50s of 220 and 310 nM for human PLD1 and PLD2, respectively. Halopemid is a dopamine receptors antagonist, and acts a psychotropic agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9515