Learn More
Apexbio Technology LLC Haloperidol 52-86-8 1g
SDP
Catalog No. 5000092310
Encompass_Preferred
Haloperidol is a small-molecule antagonist targeting dopamine receptors It is designed to block dopamine signaling thereby modulating neurotransmission within cortical limbic and nigrostriatal pathways Haloperidol exerts its biological activity primarily through inverse agonism at dopamine receptors Based on these pharmacological properties Haloperidol holds research potential in psychiatric and neuropsychological applications including studies of delirium delusions hallucinations and characterization of dopamine-mediated neuronal circuits and behavioral models in psychiatric disorders
Catalog No. 50-000-92310 Supplier Apexbio Technology LLC Supplier No. B20991000
Haloperidol is a small-molecule antagonist targeting dopamine receptors It is designed to block dopamine signaling thereby modulating neurotransmission within cortical limbic and nigrostriatal pathways Haloperidol exerts its biological activity primarily through inverse agonism at dopamine receptors Based on these pharmacological properties Haloperidol holds research potential in psychiatric and neuropsychological applications including studies of delirium delusions hallucinations and characterization of dopamine-mediated neuronal circuits and behavioral models in psychiatric disorders