Learn More
Medchemexpress LLC Haloperidol-d4-1 | 136765-35-0 | 98.0% | 379.89 g/mol | C21H19D4ClFNO2 | 5 MG
SDP
Catalog No. 5000221595
Encompass_Preferred
Haloperidol-d4-1 is a deuterium-labeled isotopologue of haloperidol intended for research and analytical applications. It is used as an internal standard and tracer for quantitative LC-MS/GC-MS and NMR studies, enabling improved accuracy in pharmacokinetic and bioanalytical assays.
  • Deuterium-labeled internal standard for LC-MS, GC-MS, and NMR quantification.
  • High purity (98.0%).
  • Improves accuracy and recovery correction in quantitative assays.
  • Solid, white to off-white form for straightforward handling.
  • Supplied in a small analytical quantity (5 mg) suitable for method development.
Catalog No. 50-002-21595 Supplier Medchemexpress LLC Supplier No. HY14538S15MG
Haloperidol-d4-1 is a deuterium-labeled isotopologue of haloperidol intended for research and analytical applications. It is used as an internal standard and tracer for quantitative LC-MS/GC-MS and NMR studies, enabling improved accuracy in pharmacokinetic and bioanalytical assays.
  • Deuterium-labeled internal standard for LC-MS, GC-MS, and NMR quantification.
  • High purity (98.0%).
  • Improves accuracy and recovery correction in quantitative assays.
  • Solid, white to off-white form for straightforward handling.
  • Supplied in a small analytical quantity (5 mg) suitable for method development.