Learn More
Medchemexpress LLC Haloperidol-d4-1 | 136765-35-0 | 98.0% | C21H19D4ClFNO2 | 1 MG
SDP
Catalog No. 5000353949
Encompass_Preferred
Haloperidol-d4-1 is a deuterium labeled version of haloperidol, which is a potent dopamine D2 receptor antagonist. Haloperidol is widely used as an antipsychotic agent and can be used in the study of various neurological and psychiatric conditions.
  • Deuterium labeled haloperidol
  • Potent dopamine D2 receptor antagonist
  • Useful for studying schizophrenia
  • Useful for studying tics in Tourette syndrome
  • Useful for studying mania in bipolar disorder
  • Useful for studying delirium
  • Useful for studying acute psychosis
  • Useful for studying hallucinations from alcohol withdrawal
  • For research use only
Catalog No. 50-003-53949 Supplier Medchemexpress LLC Supplier No. HY14538S11MG
Haloperidol-d4-1 is a deuterium labeled version of haloperidol, which is a potent dopamine D2 receptor antagonist. Haloperidol is widely used as an antipsychotic agent and can be used in the study of various neurological and psychiatric conditions.
  • Deuterium labeled haloperidol
  • Potent dopamine D2 receptor antagonist
  • Useful for studying schizophrenia
  • Useful for studying tics in Tourette syndrome
  • Useful for studying mania in bipolar disorder
  • Useful for studying delirium
  • Useful for studying acute psychosis
  • Useful for studying hallucinations from alcohol withdrawal
  • For research use only