Learn More
Medchemexpress LLC Haloperidol-d4-1 | 136765-35-0 | MFCD00673279 | 98.0% | C21H19D4ClFNO2 | 10MG
SDP
Catalog No. 5000261910
Encompass_Preferred
Haloperidol-d4-1 is a deuterium-labeled analogue of haloperidol provided as an analytical reference standard for laboratory and forensic research. It is intended for research use only and not for human or clinical use, and serves as an internal standard for quantitative mass spectrometry assays.
  • Provides a stable isotopic internal standard for quantitative LC-MS and GC-MS analysis.
  • High isotopic enrichment and consistent mass shift for accurate quantitation.
  • High purity suitable for analytical and reference applications.
  • White to off-white solid, easy to handle and store under standard conditions.
  • Available in small reference sizes compatible with method development.
  • Supplied for research use only; not for human or clinical use.
Catalog No. 50-002-61910 Supplier Medchemexpress LLC Supplier No. HY14538S110MG
Haloperidol-d4-1 is a deuterium-labeled analogue of haloperidol provided as an analytical reference standard for laboratory and forensic research. It is intended for research use only and not for human or clinical use, and serves as an internal standard for quantitative mass spectrometry assays.
  • Provides a stable isotopic internal standard for quantitative LC-MS and GC-MS analysis.
  • High isotopic enrichment and consistent mass shift for accurate quantitation.
  • High purity suitable for analytical and reference applications.
  • White to off-white solid, easy to handle and store under standard conditions.
  • Available in small reference sizes compatible with method development.
  • Supplied for research use only; not for human or clinical use.