Learn More
Medchemexpress LLC Haloperidol-d4 | 1189986-59-1 | C21H19D4ClFNO2 | 2.5 MG
SDP
Catalog No. 5000354194
Encompass_Preferred
Haloperidol-d4 is deuterium labeled haloperidol, which acts as a potent dopamine D2 receptor antagonist. This compound serves as a valuable tool in research.
  • Can be used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Potent D2 receptor antagonist.
Catalog No. 50-003-54194 Supplier Medchemexpress LLC Supplier No. HY14538S2.5MG
Haloperidol-d4 is deuterium labeled haloperidol, which acts as a potent dopamine D2 receptor antagonist. This compound serves as a valuable tool in research.
  • Can be used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Potent D2 receptor antagonist.