Learn More
Cayman Chemical HaloperIdol-d4 500ug
SDP
Catalog No. 502936745
Encompass_Preferred
An internal standard for the quantification of haloperidol by GC- or LC-MS
Catalog No. 50-293-6745 Supplier Cayman Chemical Supplier No. 26116500 UG
An internal standard for the quantification of haloperidol by GC- or LC-MS