Learn More
Medchemexpress LLC Haloperidol decanoate | 74050-97-8 | MFCD00866706 | 99.7% | 530.11 g·mol⁻¹ | C31H41ClFNO3 | 100 MG
SDP
Catalog No. 5000242239
Encompass_Preferred
Haloperidol decanoate is the decanoate ester of haloperidol supplied for research use. It is used in pharmacology and formulation studies as a long-acting depot prodrug to investigate dopamine D2 receptor activity and long-acting injectable formulations.
  • Long-acting ester for depot formulations.
  • Suitable for pharmacology and formulation studies.
  • High purity: 99.7%.
  • Molecular formula C31H41ClFNO3; molecular weight 530.11 g·mol⁻¹.
  • Available as solid packs and 10 mM DMSO solution stocks.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-42239 Supplier Medchemexpress LLC Supplier No. HY107969100MG
Haloperidol decanoate is the decanoate ester of haloperidol supplied for research use. It is used in pharmacology and formulation studies as a long-acting depot prodrug to investigate dopamine D2 receptor activity and long-acting injectable formulations.
  • Long-acting ester for depot formulations.
  • Suitable for pharmacology and formulation studies.
  • High purity: 99.7%.
  • Molecular formula C31H41ClFNO3; molecular weight 530.11 g·mol⁻¹.
  • Available as solid packs and 10 mM DMSO solution stocks.
  • For research use only; not for human or veterinary use.