Learn More
Apexbio Technology LLC Haloperidol hydrochloride 1511-16-6 200mg
SDP
Catalog No. 5000088100
Encompass_Preferred
Haloperidol hydrochloride is a small-molecule antagonist targeting dopamine D2 receptors It is designed to modulate dopaminergic neurotransmission thereby regulating neural pathways implicated in psychosis Haloperidol hydrochloride exerts its biological activity primarily through inverse agonist and antagonist effects on dopamine D2 receptors particularly within the mesocortical limbic and nigrostriatal pathways Based on these pharmacological properties haloperidol hydrochloride holds research potential in studying dopamine receptor functions dopamine-mediated neural circuits and the pathophysiological mechanisms underlying psychiatric disorders
Catalog No. 50-000-88100 Supplier Apexbio Technology LLC Supplier No. B1242200
Haloperidol hydrochloride is a small-molecule antagonist targeting dopamine D2 receptors It is designed to modulate dopaminergic neurotransmission thereby regulating neural pathways implicated in psychosis Haloperidol hydrochloride exerts its biological activity primarily through inverse agonist and antagonist effects on dopamine D2 receptors particularly within the mesocortical limbic and nigrostriatal pathways Based on these pharmacological properties haloperidol hydrochloride holds research potential in studying dopamine receptor functions dopamine-mediated neural circuits and the pathophysiological mechanisms underlying psychiatric disorders