Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Haloperidol metabolite II, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013968 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3968 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3968 Supplier MilliporeSigma Supelco Supplier No. H10225MG
Add to Cart
Add to Cart
Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Packaging Poly Bottle
CAS 34104-67-1
Physical Form Solid
Linear Formula C21H25 ClFNO2
MDL Number MFCD00055102
Synonym (+/-)-4-(4-Chlorophenyl)-alpha-(4-fluorophenyl)-4-hydroxy-1-piperidinebutanol; Reduced Haloperidol
Solubility Soluble in DMSO
Type Analytical Standard
Quantity 25 mg
Formula Weight 377.88
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.