Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Haloperidol Metabolite III, MP Biomedicals™

Catalog No. ICN15369325 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
15 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15369325 25 mg
ICN15369310 10 mg
ICN15369315 15 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15369325 Supplier MP Biomedicals, Inc Supplier No. 0215369325
Add to Cart
Add to Cart

Chemical Identifiers

CAS 366-77-8
Molecular Formula C10H9FO3
Molecular Weight (g/mol) 196.177
MDL Number MFCD00002793
InChI Key WUYWHIAAQYQKPP-UHFFFAOYSA-N
Synonym 3-4-fluorobenzoyl propionic acid, 4-4-fluorophenyl-4-oxobutanoic acid, 3-4-fluorobenzoyl propanoic acid, haloperidol metabolite iii, unii-v2z6n6jc9z, beta-4-fluorobenzoyl propionic acid, v2z6n6jc9z, 3-p-fluorobenzoyl propionic acid, 4-4-fluoro-phenyl-4-oxo-butyric acid
PubChem CID 101359
IUPAC Name 4-(4-fluorophenyl)-4-oxobutanoic acid
SMILES C1=CC(=CC=C1C(=O)CCC(=O)O)F

Specifications

Color White
Quantity 25 mg
Formula Weight 196.17
Physical Form Solid
Chemical Name or Material Haloperidol Metabolite III
EINECSNumber 206-679-1
Recommended Storage Store at Room Temperature (15° to 30°C).
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.