Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Haloxyfop-methyl | 69806-40-2 | MFCD01310644 | 99.6% | 375.73 g/mol | C16H13ClF3NO4 | 1 ML
SDP

Catalog No. 5000280766
Encompass_Preferred

Haloxyfop-methyl 10 mM (1 mL) is a research-grade solution of the aryloxyphenoxypropionic-acid herbicide haloxyfop-methyl, supplied pre-dissolved in DMSO for analytical and laboratory use. The material is intended for research and analytical applications, including qualitative and quantitative methods.

  • High purity (99.6%).
  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • Suitable for HPLC, GC, and MS analytical methods.
  • Contains CAS 69806-40-2 for unambiguous identification.
  • Molecular formula C16H13ClF3NO4; molecular weight 375.73 g/mol.

Catalog No. 50-002-80766 Supplier Medchemexpress LLC Supplier No. HYW28630710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Haloxyfop-methyl 10 mM (1 mL) is a research-grade solution of the aryloxyphenoxypropionic-acid herbicide haloxyfop-methyl, supplied pre-dissolved in DMSO for analytical and laboratory use. The material is intended for research and analytical applications, including qualitative and quantitative methods.

  • High purity (99.6%).
  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • Suitable for HPLC, GC, and MS analytical methods.
  • Contains CAS 69806-40-2 for unambiguous identification.
  • Molecular formula C16H13ClF3NO4; molecular weight 375.73 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.