Learn More
AdipoGen Haloxyfop-methyl
SDP
Supplier:  AdipoGen CDXH0230M100
Encompass_Preferred
Chemical. CAS 69806-40-2. Formula C16H13ClF3NO4. MW 375.73. Haloxyfop-methyl is a phenoxy herbicide that is used to control broadleaf weeds in crops. It is absorbed by the leaves and roots of plants and translocates to the growing points, where it interferes with the synthesis of fatty acids. This disrupts the plants cell membranes and leads to death. Compound can be used as analytical reference material.
Catalog No. 50-256-0697