Learn More
Fine Science Tools Halsey Needle Holders, Stainless Steel, 13 cm
SDP
Catalog No. NC9486852
Encompass
Product: Halsey Needle Holder - Straight/Smooth/13cm/with Lock/ Tip Shape: Straight/ Tip Width: 1.9mm/ Clamping Length: 16mm/ Scissors: No/ Lock: Yes/ Alloy: Stainless Steel/ Length: 13cm/ Serrated: No/ Unit: Each
Catalog No. NC9486852 Supplier Fine Science Tools Supplier No. 1200113
Product: Halsey Needle Holder - Straight/Smooth/13cm/with Lock/ Tip Shape: Straight/ Tip Width: 1.9mm/ Clamping Length: 16mm/ Scissors: No/ Lock: Yes/ Alloy: Stainless Steel/ Length: 13cm/ Serrated: No/ Unit: Each