Learn More
Sklar Instruments Halsey Needle Holder, Sklar, 5 in., Blunt, Straight, Cross-Serrated, Stainless Steel. EA/1.
SDP
Catalog No. 501437462 Shop All Sklar Instruments Products
Encompass_Preferred
Halsey Needle Holder, Sklar, 5 in., Blunt, Straight, Cross-Serrated, Stainless Steel. Sklar Halsey Needle Holders are used for holding small needles for suturing, most commonly during plastic surgery and procedures with delicate tissue, such as dermatology. They are available with smooth or delicately serrated jaws. Smooth jaws will cause the least damage to the needle; however, serrated jaws hold the needle more tightly. Physician preference typically determines the type of working surface utilized. This product is straight with cross-serrated tips and a length of 5 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7462 Supplier Sklar Instruments Supplier No. 201751
Halsey Needle Holder, Sklar, 5 in., Blunt, Straight, Cross-Serrated, Stainless Steel. Sklar Halsey Needle Holders are used for holding small needles for suturing, most commonly during plastic surgery and procedures with delicate tissue, such as dermatology. They are available with smooth or delicately serrated jaws. Smooth jaws will cause the least damage to the needle; however, serrated jaws hold the needle more tightly. Physician preference typically determines the type of working surface utilized. This product is straight with cross-serrated tips and a length of 5 inches.