Learn More
Sklar Instruments Surgi-OR TC Halsey Needle Holder, Sklar 5 in., Cross-Serrated, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. EA.
SDP
Catalog No. 501184167 Shop All Sklar Instruments Products
Encompass_Preferred
Surgi-OR TC Halsey Needle Holder, Sklar. 5 in., Cross-Serrated, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. Surgi-OR TC Halsey Needle Holders are used for holding small needles for suturing, most commonly during plastic surgery and procedures with delicate tissue, such as dermatology. They are available with smooth or delicately serrated jaws. Smooth jaws will cause the least damage to the needle; however, serrated jaws hold the needle more tightly. Physician preference typically determines the type of working surface utilized. Surgi-OR TC mid-grade products are reliable instrumentation for the operating room and other surgical centers. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. This product is straight with cross-serrated tips and a length of 5 inches.
Catalog No. 50-118-4167 Supplier Sklar Instruments Supplier No. 95133
Surgi-OR TC Halsey Needle Holder, Sklar. 5 in., Cross-Serrated, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. Surgi-OR TC Halsey Needle Holders are used for holding small needles for suturing, most commonly during plastic surgery and procedures with delicate tissue, such as dermatology. They are available with smooth or delicately serrated jaws. Smooth jaws will cause the least damage to the needle; however, serrated jaws hold the needle more tightly. Physician preference typically determines the type of working surface utilized. Surgi-OR TC mid-grade products are reliable instrumentation for the operating room and other surgical centers. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. This product is straight with cross-serrated tips and a length of 5 inches.