Learn More
Fine Science Tools Halsey Micro Needle Holder, Stainless Steel, 12.5 cm
SDP
Catalog No. NC9903046
Encompass
Product: Halsey Micro Needle Holder/ Tip Shape: Straight/ Tip Width: 1.4mm/ Clamping Length: 12.5mm/ Scissors: No/ Lock: Yes/ Alloy: Stainless Steel/ Length: 12.5cm/ Serrated: Yes/ Feature: Tungsten Carbide/ Unit: Each
Catalog No. NC9903046 Supplier Fine Science Tools Supplier No. 1250012
Product: Halsey Micro Needle Holder/ Tip Shape: Straight/ Tip Width: 1.4mm/ Clamping Length: 12.5mm/ Scissors: No/ Lock: Yes/ Alloy: Stainless Steel/ Length: 12.5cm/ Serrated: Yes/ Feature: Tungsten Carbide/ Unit: Each