Learn More
Electron Microscopy Sciences EMS Halstead Mosquito Forceps Standard, Curved
SDP
Catalog No. 501931467
Encompass_Preferred
The forceps have a finger ring grip and adjustable interlocking cross arms that offers secure clamping.
Blade: Curved
OAL: 5"
This item is not returnable. View return policy
Catalog No. 50-193-1467 Supplier Electron Microscopy Sciences Supplier No. 7299924
The forceps have a finger ring grip and adjustable interlocking cross arms that offers secure clamping.
Blade: Curved
OAL: 5"