Learn More
Stoelting™ Halstead Mosquito Forceps
SDP
Catalog No. 10000476 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Curved
Curved, Delicate
Straight
Straight, Delicate
Straight, Left-handed
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-000-476 Curved, Delicate
10-000-473 Straight
10-000-472 Straight, Left-handed
10-000-475 Straight, Delicate
10-000-474 Curved
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 10-000-476 Supplier Stoelting™ Supplier No. 5211013P
Add to Cart
Add to Cart
Hemostatic forceps are used in many surgical procedures usually to control bleeding or the flow of liquids in tubing.
  • Scissor-like ring handles
  • Ratcheted handles can be locked in multiple positions in order to maintain variable levels of constant pressure.
  • Delicate hemostats have thin jaws with fine serrations
Specifications
For Use With (Application) Neuroscience
Length (English) 5 in.
Model Curved, Delicate