Learn More
Electron Microscopy Sciences Halstead Mosquito Forcept, Curved, 127 mm OAL
SDP
Catalog No. 5026421
Encompass_Preferred
Stainless Steel hemostatic forceps with box lock joint and serrated jaws.
OAL: 127 mm
Catalog No. 50-264-21 Supplier Electron Microscopy Sciences Supplier No. 729425C
Stainless Steel hemostatic forceps with box lock joint and serrated jaws.
OAL: 127 mm