Learn More
Roboz Surgical HALSTED MOSQUITO 5IN CURVE
SDP
Catalog No. NC9763456
Encompass
Halstead Mosquito Forceps; Curved; 5 in. Length, 1.3mm tip diameter, 2.1mm jaw width The Halstead Mosquito Forceps are our standard fine curved hemostats. The tip diameter is 1.3mm. The jaw width is 2.1mm at 5mm from the tip.
Catalog No. NC9763456 Supplier Roboz Surgical Supplier No. RS7111
Halstead Mosquito Forceps; Curved; 5 in. Length, 1.3mm tip diameter, 2.1mm jaw width The Halstead Mosquito Forceps are our standard fine curved hemostats. The tip diameter is 1.3mm. The jaw width is 2.1mm at 5mm from the tip.