Learn More
Fine Science Tools Halsted-Mosquito Hemostats, Stainless Steel, 12.5 cm
SDP
Catalog No. NC9735639
Encompass
Tip Shape: Curved/ Tips: Serrated/ Tip Width: 1.1mm/ Clamping Length: 20mm/ Alloy / Material: Stainless Steel/ Length: 12.5cm
Catalog No. NC9735639 Supplier Fine Science Tools Supplier No. 1300912
Tip Shape: Curved/ Tips: Serrated/ Tip Width: 1.1mm/ Clamping Length: 20mm/ Alloy / Material: Stainless Steel/ Length: 12.5cm