Learn More
Sklar Instruments Econo Halsted Mosquito Forceps, 5 in., Blunt, Straight, Serrated, Reusable. BX12.
SDP
Catalog No. 501109753 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Halsted Mosquito Forceps, 5 in., Blunt, Straight, Serrated, Reusable. Econo Halsted Mosquito Forceps are used in multiple procedures as a hemostat for small blood vessels. The ratcheted finger ring handle provides a secure, locking grip to halt blood flow. They are not appropriate for use with large vessels or arteries. A toothed variation is available for grasping delicate tissue in skin grafting, biopsies, or ophthalmologic procedures. Mosquito forceps are known for their fine tips, and short, fully serrated jaws. They are available curved or straight. The standard length for this instrument is 5 inches, although Sklar does offer some with a length of 5-1/2 inches. Halsted Mosquito Forceps are very similar to Hartmann Mosquito Forceps, however, the Hartmann style is finer and more delicate. This product is straight with serrated tips and a length of 5 inches. They are sold in boxes of 12.
Catalog No. 50-110-9753 Supplier Sklar Instruments Supplier No. 21426
Econo Halsted Mosquito Forceps, 5 in., Blunt, Straight, Serrated, Reusable. Econo Halsted Mosquito Forceps are used in multiple procedures as a hemostat for small blood vessels. The ratcheted finger ring handle provides a secure, locking grip to halt blood flow. They are not appropriate for use with large vessels or arteries. A toothed variation is available for grasping delicate tissue in skin grafting, biopsies, or ophthalmologic procedures. Mosquito forceps are known for their fine tips, and short, fully serrated jaws. They are available curved or straight. The standard length for this instrument is 5 inches, although Sklar does offer some with a length of 5-1/2 inches. Halsted Mosquito Forceps are very similar to Hartmann Mosquito Forceps, however, the Hartmann style is finer and more delicate. This product is straight with serrated tips and a length of 5 inches. They are sold in boxes of 12.