Learn More
Roboz Surgical HALSTED MOSQUITO HEMOS.FORCEP
SDP
Supplier:  Roboz Surgical RS7110
Encompass
Halstead Mosquito Forceps; Straight; 5 in. Length, 1.3mm tip diameter, 2.1mm jaw width The Halstead Mosquito Forceps are our standard fine straight hemostats. The tip diameter is 1.3mm. The jaw width is 2.1mm at 5mm from the tip.
Catalog No. NC9451354