Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Halt™ Phosphatase Inhibitor Single-Use Cocktail
GSA_VA

Catalog No. PI78427 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
10 mL
24 x 100 μL
5 x 1 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI78427 10 mL
PI78420 1 mL
PI78428 24 x 100 μL
PI78426 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PI78427 Supplier Thermo Scientific™ Supplier No. 78427
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stabilize serine, threonine and tyrosine phosphorylation of proteins in cells and tissues with this cocktail of phosphatase inhibitors.

Thermo Scientific Halt Phosphatase Inhibitor Single Use Cocktail preserves the phosphorylation state of proteins during and after cell lysis or tissue protein extraction.

Features of Halt Phosphatase Inhibitor Single Use Cocktail:

  • Devoid of protease inhibitors—To protect against both proteases and phosphatases, use Halt Protease and Phosphatase Inhibitor Cocktail (Cat. No. 78420)
  • Dialyzable— Either dialyze or desalt sample to effectively remove inhibitors from cell extracts before performing procedures that are discovered to be incompatible with cocktail components
  • Supplied as a 100X concentrate—effective when used at a final concentration of 1X; however, samples that contain particularly high levels of phosphatase activity may require a final concentration of 2-3X
  • Compatibility— compatible with most detergent-based cell lysis reagents, including all Thermo Scientific Pierce Cell Lysis Solutions
  • All-in-one format—inhibits both serine-threonine phosphatases and protein tyrosine phosphatases
  • Compatible with popular protein assays—works with standard protein assays, including BCA and coomassie (Bradford) assays
  • Convenient storage—refrigerator (2 to 8°C) storage saves valuable space in the freezer and provides for immediate use

Halt Phosphatase Inhibitor Cocktail protects phosphoproteins from serine/threonine phosphatases as well as protein tyrosine phosphatases (PTPs) during cell lysis. The cocktail contains a mixture of four inhibitors of broad specificity, including sodium fluoride, sodium orthovanadate, sodium pyrophosphate and beta-glycerophosphate. Conveniently packaged in a 100X format, the cocktail is stable at 4°C and is suitable for use with any cell or tissue extract. The cocktail preserves protein phosphorylation in immunoprecipitation and kinase assays.

Phosphorylation and dephosphorylation comprise a molecular on/off switch that regulates many key biological pathways within the cell, including signal transduction, cell division and apoptosis. To effectively study phosphorylation activation states, one must inactivate phosphatases to prevent their uncontrolled activity upon cell lysis. Halt Phosphatase Inhibitor Cocktail accomplishes this task.

Related Products

  • Pierce™ Phosphatase Inhibitor Mini Tablets (Cat. No. 88667)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Phosphatase Inhibitor Cocktail
Content And Storage Upon receipt store product at 4°C. Do not freeze.
Form Liquid
Product Line Halt
Reagent Type Phosphatase Inhibitor
Inhibitors Phosphatase Inhibitor
Quantity 10 mL
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.