Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Halt™ Protease and Phosphatase Inhibitor Cocktail, EDTA-free (100X)
GSA_VA

Catalog No. PI78445 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
10 mL
24 x 100 μL
5 x 1 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI78445 5 x 1 mL
PI78441 1 mL
PI78443 24 x 100 μL
PI78447 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PI78445 Supplier Thermo Scientific™ Supplier No. 78445
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Prevent protein degradation and stabilize phosphorylation with this all-in-one cocktail of protease and phosphatase inhibitors.

The all-in-one Thermo Scientific™ Halt Protease and Phosphatase Inhibitor Cocktail provides the convenience of a single solution with full sample protection.

This broad-spectrum Halt Protease and Phosphatase Inhibitor Cocktail is specifically optimized to protect proteins from degradation during extraction and purification. The cocktail contains inhibitors against the major classes of proteases and phosphatases, targeting aminopeptidases, cysteine and serine proteases and serine/threonine and protein tyrosine phosphatases. The 100X cocktails, available in regular and EDTA-free formulations and several package formats, are refrigerator-stable liquids that are very easy to use for nearly any sample or experimental scale.

Highlights:

  • Complete protection – all-in-one cocktail contains both protease and phosphatase inhibitors
  • Multiple package sizes – choose from four convenient packages sizes to match nearly any scale
  • Convenient – the 100X liquid formulation is much easier and more effective to use than individual inhibitors and other formats
  • Easy storage – aqueous-based format allows for convenient cold-room storage without freezing, by contrast with DMSO-based cocktails
  • Compatible – can be used in mass spectrometry (MS) because it does not contain AEBSF, which can cause peaks to shift
  • Validated – fully compatible with Thermo Scientific Pierce™ Cell Lysis Buffers

Includes:

Formulations with and without supplementary vial of EDTA for metalloprotease inhibition, both of which are offered in four package sizes.

Storage Requirements:

Upon receipt store at 4°C. Do not freeze product.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Halt Protease and Phosphatase Inhibitor Cocktail, EDTA-Free
Product Type Protease and Phosphatase Inhibitor Cocktail
Content And Storage Upon receipt store at 4°C. Product shipped with an ice pack. Do not freeze product.
Form Liquid
Product Line Halt
Buffer Lysis Buffer
Reagent Type Protease Inhibitor, Phosphatase Inhibitor
Inhibitors Protease Inhibitor, Phosphatase Inhibitor
Quantity 5 x 1 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.