Learn More
CAS DATALOGGERS Halveon T Stream Cryogenic Single Use PDF Temperature Data Logger for -200C Goods
Catalog No. NC3847099
Encompass_Preferred
"Introducing the T-Stream Cryogenic SU: a cutting-edge instrument designed to deliver accurate and reliable temperature monitoring in the most challenging environments at an affordable price. With an external probe, this device can measure down to -200C to monitor your most critical goods during transit.Engineered having the probe directly in contact with -200C contents, this data logger offers exceptional durability and precision. The logger has a unique button design that prevents accidental stops during transit and ensures a complete report for every temperature monitoring task. A built-in LCD screen allows you to directly check the temperature statistics, alarm status, running status, logging duration, and mark information."
Catalog No. NC3847099 Supplier CAS DATALOGGERS Supplier No. TSTREAMCRYOSU
"Introducing the T-Stream Cryogenic SU: a cutting-edge instrument designed to deliver accurate and reliable temperature monitoring in the most challenging environments at an affordable price. With an external probe, this device can measure down to -200C to monitor your most critical goods during transit.Engineered having the probe directly in contact with -200C contents, this data logger offers exceptional durability and precision. The logger has a unique button design that prevents accidental stops during transit and ensures a complete report for every temperature monitoring task. A built-in LCD screen allows you to directly check the temperature statistics, alarm status, running status, logging duration, and mark information."