Learn More
CAS DATALOGGERS Halveon T Stream Dry Ice Single Use Temperature PDF Data Logger
Catalog No. NC3847095
Encompass_Preferred
"Introducing the T-Stream Dry Ice SU: a cutting-edge instrument designed to deliver accurate and reliable temperature and humidity monitoring in the most challenging environments at an affordable price. The device is waterproof and can measure between a range of -90C to 70C.Engineered for direct immersion in dry ice, this data logger offers exceptional durability and precision. The logger has a unique button design that prevents accidental stops during transit and ensures a complete report for every temperature monitoring task. A built-in LCD screen allows you to directly check the temperature statistics, alarm status, running status, logging duration, and mark information."
Catalog No. NC3847095 Supplier CAS DATALOGGERS Supplier No. TSTREAMDRYICESU
"Introducing the T-Stream Dry Ice SU: a cutting-edge instrument designed to deliver accurate and reliable temperature and humidity monitoring in the most challenging environments at an affordable price. The device is waterproof and can measure between a range of -90C to 70C.Engineered for direct immersion in dry ice, this data logger offers exceptional durability and precision. The logger has a unique button design that prevents accidental stops during transit and ensures a complete report for every temperature monitoring task. A built-in LCD screen allows you to directly check the temperature statistics, alarm status, running status, logging duration, and mark information."