Learn More
CAS DATALOGGERS Halveon T Stream Multi Use PDF Temperature Data Logger
Catalog No. NC3847093
Encompass_Preferred
The T-Stream MU is a temperature monitoring device with a six point calibration certificate onboard and equipped with an innovative, high accuracy sensor. The device has a 2 year battery life. Its temperature range goes from -30°C ~ +70°C. Users can easily access detailed data reports in PDF and CSV format. The large LCD display makes it simple to read and accurately monitor temperature and humidity readings.
Catalog No. NC3847093 Supplier CAS DATALOGGERS Supplier No. TSTREAMMULTIUSE5X
The T-Stream MU is a temperature monitoring device with a six point calibration certificate onboard and equipped with an innovative, high accuracy sensor. The device has a 2 year battery life. Its temperature range goes from -30°C ~ +70°C. Users can easily access detailed data reports in PDF and CSV format. The large LCD display makes it simple to read and accurately monitor temperature and humidity readings.