Learn More
CAS DATALOGGERS Halveon T Stream Single Use PDF Temperature Data Logger
Catalog No. NC3847092
Encompass_Preferred
The T-Stream SU is a temperature monitoring device with a six point calibration certificate onboard and equipped with an innovative, high accuracy sensor. Its temperature range goes from -30°C ~ +70°C and its packaging is waterproof. Users can easily access detailed data reports in PDF and CSV format. The large LCD display makes it simple to read and accurately monitor temperature and humidity readings.
Catalog No. NC3847092 Supplier CAS DATALOGGERS Supplier No. TSTREAMSINGLEUSE5X
The T-Stream SU is a temperature monitoring device with a six point calibration certificate onboard and equipped with an innovative, high accuracy sensor. Its temperature range goes from -30°C ~ +70°C and its packaging is waterproof. Users can easily access detailed data reports in PDF and CSV format. The large LCD display makes it simple to read and accurately monitor temperature and humidity readings.