Learn More
Halyard Health BASIC™ Plus Lab Coats with Knit Collar and Cuff
Catalog No. 23500161
Change view
Click to view available options
Color:
Blue
White
Size:
Small
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
23-500-161 White Large
23-500-162 White X-Large
23-500-163 White 2X-Large
23-500-164 Blue Small
23-500-166 Blue Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 23-500-161 Supplier Halyard Health Supplier No. 10022
Add to Cart
Add to Cart
This item is not returnable. View return policy
Protective tri-layer fabric offers resistance to blood strikethrough at various levels of exposure.
  • For low exposure to fluids or chemicals
  • Non-sterile
  • Knit collar and cuffs
  • Five snaps
  • Three reinforced pockets
  • Neck loop for hanging
  • Size/name label
  • Available in colors blue and white
Specifications
Product Type General Purpose Lab Coat
Size Large
Gender Unisex
Material SMS
Color White
Cuff Style Knit
Size (Metric) Chest 67.75 cm
Length (Metric) 115.6 cm
Construction 3-ply SMS