Learn More
Halyard Health BASIC™ Plus Lab Coats with Knit Collar and Cuff
Catalog No. 23500166
Change view
Click to view available options
Color:
Blue
White
Size:
Small
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
23-500-166 Blue Large
23-500-161 White Large
23-500-162 White X-Large
23-500-163 White 2X-Large
23-500-164 Blue Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 23-500-166 Supplier Halyard Health Supplier No. 10032
Add to Cart
Add to Cart
This item is not returnable. View return policy
Protective tri-layer fabric offers resistance to blood strikethrough at various levels of exposure.
  • For low exposure to fluids or chemicals
  • Non-sterile
  • Knit collar and cuffs
  • Five snaps
  • Three reinforced pockets
  • Neck loop for hanging
  • Size/name label
  • Available in colors blue and white
Specifications
Product Type General Purpose Lab Coat
Size Large
Gender Unisex
Material SMS
Color Blue
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
Resistance Fluids
No. of Pockets 3
Size (Metric) Chest 67.75 cm
Length (Metric) 115.6 cm
Construction 3-ply SMS