Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc Hams F-12 without Hypoxanthine and L-Glutamine, 1000 mL
SDP

Catalog No. 507517657
Encompass_Preferred

Compared to other basal media, Hams F-12 contains a wider variety of components, and no proteins or growth factors

Without hypoxanthine and L-glutamine
Applications: Useful for serum free clonal growth of Chinese Hamster Ovary (CHO) cells, chick embryos and mouse L-cells Also used to culture myeloma cells and hybridomas, primary rat hepatocytes and prostate epithelial cells

Storage Conditions: 2 °C to 8 °C
Shipping Condition: ambient temperature
Sterile Filtered Osmolality: 295 ± 5% mOsm/Kg H20 Max Shelf Life: 24 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-751-7657 Supplier Quality Biological Inc Supplier No. 112153131
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compared to other basal media, Hams F-12 contains a wider variety of components, and no proteins or growth factors

Without hypoxanthine and L-glutamine
Applications: Useful for serum free clonal growth of Chinese Hamster Ovary (CHO) cells, chick embryos and mouse L-cells Also used to culture myeloma cells and hybridomas, primary rat hepatocytes and prostate epithelial cells

Storage Conditions: 2 °C to 8 °C
Shipping Condition: ambient temperature
Sterile Filtered Osmolality: 295 ± 5% mOsm/Kg H20 Max Shelf Life: 24 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.