Learn More
MOLECULAR DEPOT LLC HAMA 1000-1499 ng/mL - 2/1 mL
Catalog No. 5000013556
Encompass_Preferred
High HAMA Serum and Plasma Matched Set (1000-1499 ng/mL) is a matched set of Serum and K3 EDTA Plasma samples (1 mL each) containing human anti-mouse antibodies (HAMA) Titers are established by a cleared HAMA ELISA method These samples are to be used to test HAMA interference in immunoassays For in vitro research use only
Catalog No. 50-000-13556 Supplier MOLECULAR DEPOT LLC Supplier No. S2010006P1
High HAMA Serum and Plasma Matched Set (1000-1499 ng/mL) is a matched set of Serum and K3 EDTA Plasma samples (1 mL each) containing human anti-mouse antibodies (HAMA) Titers are established by a cleared HAMA ELISA method These samples are to be used to test HAMA interference in immunoassays For in vitro research use only