Learn More
MOLECULAR DEPOT LLC HAMA 800-999 ng/mL - 1 mL
Supplier:  MOLECULAR DEPOT LLC S2010006B
Encompass_Preferred
High HAMA Serum (800-999 ng/mL) is a serum sample containing human anti-mouse antibodies (HAMA) with a titer established by a cleared HAMA ELISA method This sample is to be used to test HAMA interference in immunoassays For in vitro research use only
Catalog No. 50-000-13325