Learn More
MOLECULAR DEPOT LLC HAMA 800-999 ng/mL - 1 mL
Catalog No. 5000013552
Encompass_Preferred
High HAMA Li-Heparin Plasma (800-999 ng/mL) is an Li-Heparin plasma sample containing human anti-mouse antibodies (HAMA) with a titer established by a cleared HAMA ELISA method This sample is to be used to test HAMA interference in immunoassays For in vitro research use only
Catalog No. 50-000-13552 Supplier MOLECULAR DEPOT LLC Supplier No. S2010006P2B
High HAMA Li-Heparin Plasma (800-999 ng/mL) is an Li-Heparin plasma sample containing human anti-mouse antibodies (HAMA) with a titer established by a cleared HAMA ELISA method This sample is to be used to test HAMA interference in immunoassays For in vitro research use only