Learn More
MOLECULAR DEPOT LLC HAMA Blocking Reagent - 10 mg
Catalog No. 5000013503
Encompass_Preferred
HAMA Blocking Reagent (Affinity Purified) consists of a proprietary blend of proteins that specifically binds to HAMA minimizing their interference and significantly reducing the number of false positives To reduce HAMA interference simply spike our HAMA Blocking Reagent into your immunoassay reagent(s) to 5-50 ug/mL (optimum concentration is assay dependent) For in vitro research use only
Catalog No. 50-000-13503 Supplier MOLECULAR DEPOT LLC Supplier No. A2010010
Add to Cart
Add to Cart
HAMA Blocking Reagent (Affinity Purified) consists of a proprietary blend of proteins that specifically binds to HAMA minimizing their interference and significantly reducing the number of false positives To reduce HAMA interference simply spike our HAMA Blocking Reagent into your immunoassay reagent(s) to 5-50 ug/mL (optimum concentration is assay dependent) For in vitro research use only