Learn More
Carbosynth LLC. HAMA blocking reagent, 1 gram
SDP
Catalog No. 501870913
Encompass_Preferred
HAMA Blocking Reagent for use in immunoassays such as ELISA
Catalog No. 50-187-0913 Supplier Carbosynth LLC. Supplier No. 85R1001P0.05G
HAMA Blocking Reagent for use in immunoassays such as ELISA