Learn More
MOLECULAR DEPOT LLC HAMA Interference Kit - 6/2 mL
Catalog No. 5000013443
Encompass_Preferred
HAMA Interference Kit consists of six serum samples (2 mL each) that can be used to test HAMA interference in immunoassays The kit includes five HAMA-positive serum samples (PS1 to PS5) containing HAMA concentrations of up to 200 ng/mL and one HAMA-negative serum sample (NS) HAMA titers were established by cleared HAMA ELISA method Instructions with sample calculations are included For in vitro research use only
Catalog No. 50-000-13443 Supplier MOLECULAR DEPOT LLC Supplier No. K2010009
HAMA Interference Kit consists of six serum samples (2 mL each) that can be used to test HAMA interference in immunoassays The kit includes five HAMA-positive serum samples (PS1 to PS5) containing HAMA concentrations of up to 200 ng/mL and one HAMA-negative serum sample (NS) HAMA titers were established by cleared HAMA ELISA method Instructions with sample calculations are included For in vitro research use only