Learn More
MOLECULAR DEPOT LLC HAMA over 10000 ng/mL - 1 mL
Catalog No. 5000013346
Encompass_Preferred
High HAMA K3 EDTA Plasma (over 100000 ng/mL) is a K3 EDTA plasma sample containing human anti-mouse antibodies (HAMA) with a titer established by a cleared HAMA ELISA method This sample is to be used to test HAMA interference in immunoassays For in vitro research use only
Catalog No. 50-000-13346 Supplier MOLECULAR DEPOT LLC Supplier No. S2010006P1I
Add to Cart
Add to Cart
Request a Quote
High HAMA K3 EDTA Plasma (over 100000 ng/mL) is a K3 EDTA plasma sample containing human anti-mouse antibodies (HAMA) with a titer established by a cleared HAMA ELISA method This sample is to be used to test HAMA interference in immunoassays For in vitro research use only