Learn More
TARGETMOL CHEMICALS INC HAMAMELITANNIN 5MG
SDP
Catalog No. 502578890
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Hamamelitannin a polyphenol extracted from the bark of Hamamelis virginiana has cytotoxic and antibiofilm activities,and is a quorum-sensing (QS) inhibitor.It increases antibiotic susceptibility of staphylococcus aureus biofilms by affecting peptidoglycan biosynthesis and eDNA release.It also has a high protective activity on cell damage induced by peroxides. purity: 99%
Catalog No. 50-257-8890 Supplier TARGETMOL CHEMICALS INC Supplier No. TN17225MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Hamamelitannin a polyphenol extracted from the bark of Hamamelis virginiana has cytotoxic and antibiofilm activities,and is a quorum-sensing (QS) inhibitor.It increases antibiotic susceptibility of staphylococcus aureus biofilms by affecting peptidoglycan biosynthesis and eDNA release.It also has a high protective activity on cell damage induced by peroxides. purity: 99%