Learn More
Cell Signaling Technology Hamartin/TSC1 Monoclonal Antibody, Rabbit, Unconjugated, 20 μL
SDP
Catalog No. 502805256
Encompass
The Hamartin/TSC1 (D43E2) Rabbit Monoclonal Antibody is designed to detect endogenous levels of total TSC1 protein. This monoclonal antibody is produced by immunizing animals with a synthetic peptide corresponding to residues surrounding Val640 of human TSC1. The TSC1 protein, also known as hamartin, functions as a tumor suppressor and forms a critical complex with tuberin to regulate cellular processes such as mTOR signaling, cell cycle control, and vesicular trafficking.
  • Validated for use in Western Blotting and Immunoprecipitation.
  • Species reactivity includes human, mouse, and rat.
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide.
  • Recommended dilution for Western Blotting is 1:1,000.
  • Recommended dilution for Immunoprecipitation is 1:50.
  • Formulated for storage at −20°C.
Catalog No. 50-280-5256 Supplier Cell Signaling Technology Supplier No. 6935T
The Hamartin/TSC1 (D43E2) Rabbit Monoclonal Antibody is designed to detect endogenous levels of total TSC1 protein. This monoclonal antibody is produced by immunizing animals with a synthetic peptide corresponding to residues surrounding Val640 of human TSC1. The TSC1 protein, also known as hamartin, functions as a tumor suppressor and forms a critical complex with tuberin to regulate cellular processes such as mTOR signaling, cell cycle control, and vesicular trafficking.
  • Validated for use in Western Blotting and Immunoprecipitation.
  • Species reactivity includes human, mouse, and rat.
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide.
  • Recommended dilution for Western Blotting is 1:1,000.
  • Recommended dilution for Immunoprecipitation is 1:50.
  • Formulated for storage at −20°C.