Learn More
MSC Hamilton Caster Rigid Caster, 6x2 inch Ergo-Glide Wheel, 960 lbs Capacity
Catalog No. 5000534639
Encompass
This rigid caster is engineered for heavy-duty applications, featuring a 6x2 inch Ergo-Glide wheel with a 960 lbs load capacity. The Ergo-Glide wheel, composed of polyurethane on an iron core, provides durability and smooth operation. Its robust forged steel construction is designed for demanding industrial environments.
  • Designed for demanding industrial applications.
  • Features a 1/4 inch thick drop forged steel mounting plate and robotically welded steel legs.
  • Sturdy 3/4 inch diameter kingpin is integrally forged with the mounting plate.
  • Equipped with a CNC-machined 2 9/16 inch diameter raceway and precision tapered thrust bearing.
  • Powder coated caster rigs provide premium durability.
  • Includes ball-check type lubrication fittings.
  • Utilizes precision sealed ball bearings.
Catalog No. 50-005-34639 Supplier MSC Supplier No. 75012948
This rigid caster is engineered for heavy-duty applications, featuring a 6x2 inch Ergo-Glide wheel with a 960 lbs load capacity. The Ergo-Glide wheel, composed of polyurethane on an iron core, provides durability and smooth operation. Its robust forged steel construction is designed for demanding industrial environments.
  • Designed for demanding industrial applications.
  • Features a 1/4 inch thick drop forged steel mounting plate and robotically welded steel legs.
  • Sturdy 3/4 inch diameter kingpin is integrally forged with the mounting plate.
  • Equipped with a CNC-machined 2 9/16 inch diameter raceway and precision tapered thrust bearing.
  • Powder coated caster rigs provide premium durability.
  • Includes ball-check type lubrication fittings.
  • Utilizes precision sealed ball bearings.