Learn More
MSC Hamilton Caster, Swivel, Phenolic, 10 in Wheel Diameter, 2,900 lb Capacity
Catalog No. 5000530991
Encompass
This top plate caster features a phenolic wheel with a 10 in diameter and 3 in width, designed for demanding manufacturing applications. It offers a robust 2,900 lb load capacity and incorporates a hot forged kingpin swivel construction for reliable performance.
  • Plate mount type
  • Phenolic wheel material
  • 10 in wheel diameter
  • 3 in wheel width
  • 2,900 lb load capacity
  • Black wheel color
  • Roller bearing type
  • Hot forged swivel construction
  • Kingpin swivel design
  • 12-1/2 in overall height
  • Enamel finish
  • High relative capacity
  • Medium impact-resistant
  • Medium abrasion-resistant
Catalog No. 50-005-30991 Supplier MSC Supplier No. 75013037
This top plate caster features a phenolic wheel with a 10 in diameter and 3 in width, designed for demanding manufacturing applications. It offers a robust 2,900 lb load capacity and incorporates a hot forged kingpin swivel construction for reliable performance.
  • Plate mount type
  • Phenolic wheel material
  • 10 in wheel diameter
  • 3 in wheel width
  • 2,900 lb load capacity
  • Black wheel color
  • Roller bearing type
  • Hot forged swivel construction
  • Kingpin swivel design
  • 12-1/2 in overall height
  • Enamel finish
  • High relative capacity
  • Medium impact-resistant
  • Medium abrasion-resistant