Learn More
MSC Hamilton Caster Wheel, Polyurethane On Polypropylene, 5 In Dia, 1-3/8 In Wide
Catalog No. 5000528207
Encompass
The Hamilton Caster Wheel is constructed from polyurethane on polypropylene, featuring a 5-in diameter and a 1-3/8 in width. It includes a ball bearing for smooth operation and a 3/8 in axle. This wheel is designed for demanding applications, offering resistance to high temperatures, chemicals, and abrasion.
  • High temperature resistance (up to 180°F)
  • Quiet operation
  • Moisture-resistant
  • Rolling ease
  • Chemical-resistant
  • Abrasion-resistant
  • Floor protection
Catalog No. 50-005-28207 Supplier MSC Supplier No. 52129111
The Hamilton Caster Wheel is constructed from polyurethane on polypropylene, featuring a 5-in diameter and a 1-3/8 in width. It includes a ball bearing for smooth operation and a 3/8 in axle. This wheel is designed for demanding applications, offering resistance to high temperatures, chemicals, and abrasion.
  • High temperature resistance (up to 180°F)
  • Quiet operation
  • Moisture-resistant
  • Rolling ease
  • Chemical-resistant
  • Abrasion-resistant
  • Floor protection