Learn More
Hamilton™ Chemically Inert Manual Valves
Catalog No. 14815354 Shop All Hamilton Company Products
Change view
Click to view available options
Type:
1-1 Manual Valve
3-2 Manual Valve
3-3 Manual Valve
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
14-815-354 3-3 Manual Valve
14-819-280 1-1 Manual Valve
14-819-281 3-2 Manual Valve
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-815-354 Supplier Hamilton™ Supplier No. 86727
Add to Cart
Add to Cart
Valves have a fluid path containing PTFE and CTFE
The ports on the valve are 1/4-28 threads and come in different flow paths.
Specifications
For Use With (Application) For low pressure applications, from a basic on/off flow to a four-way distribution system
Material CTFE, PTFE
Type 3-3 Manual Valve