Description
- Aluminum housing with PTFE-rotary plug
- Rotation of valve connects two syringes to same fluid path
- Enables larger dilutions
Specifications
Specifications
| For Use With (Equipment) | ML503 A and ML530 B Instruments |
| Material | Aluminum, PTFE |
| Pressure | 0 to 100 psi |
| For Use With (Application) | Rotation of Valve Connects Two Syringes to Same Fluid Path |
| Product Type | Dual Syringe Diluter Valve |