Learn More
Hamilton™ Disposable Tip Adapter
Catalog No. 1368478F Shop All Hamilton Company Products
Change view
Click to view available options
Volume (Metric):
100 μL
Catalog No. Volume (Metric)
13-684-78F 100 μL
1 options
Catalog No. 13-684-78F Supplier Hamilton™ Supplier No. 31330
Add to Cart
Edge
Add to Cart
Designed to enable the connection of a standard 1mL disposable pipette tip to the end of a Hamilton™ syringe.
  • Adapter connects to standard Luer Tip (LT) and PTFE Luer Lock (TLL) syringes
  • Ideal for eliminating the possibility for cross-contamination
  • Manufactured from CTFE
  • Provided individually
Specifications
Material CTFE
Product Type Luer-tip Adapter
Volume (Metric) 100 μL
For Use With (Equipment) PB600-1 Repeating Dispensers